Stearic acid
Catalog # EHERC15710120
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:284.48 g/mol
Merck Index:13,08882
Boiling Pt:361 °C (1013 hPa)
Melting Pt:67 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:196 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:200-313-4
CAS Number:57-11-4
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00002752