Stearic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:57-11-4
:
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00002752
:284.48 g/mol
Merck Index:13,08882
Boiling Pt:361 °C (760 mmHg)
Melting Pt:67…69 °C
Density:0.84 g/cm³
Flash Pt:196 °C
Storage Temperature:Ambient
EINECS:200-313-4
Specification Test Results
- Appearance:White Granular powder or granules or flakes
- Infrared spectrum:Conforms
- Melting point:67 to 71 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together