Order Entry
Belgium
ContactUsLinkComponent
Formic acid ≥99%
Formic acid ≥99%
Catalog # PIER28905
CAS Number:  
undefined
Formic acid ≥99%
Catalog # PIER28905
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Seq:
    0125

Specifications

About this item

 :
 :46.03 g/mol
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8,II
 :GHS05
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297