Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Formic acid ≥98%, GPR RECTAPUR®
 
undefined
 
 
Formic acid ≥98%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 98 %
  • Refractive index (20°C):
    1.370 - 1.372
  • Acetic acid:
    Max. 0.2 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 100 ppm
  • Evaporation residue:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 50 ppm

 

Frequently Bought Together