Order Entry
Belgium
ContactUsLinkComponent
Formic acid ≥88.0% (by titrimetric analysis), TraceSELECT™ for trace analysis, Fluka™
 
 
 
Formic acid ≥88.0% (by titrimetric analysis), TraceSELECT™ for trace analysis, Fluka™
  06454-250ML
 :  
undefined
 
 
 
Formic acid ≥88.0% (by titrimetric analysis), TraceSELECT™ for trace analysis, Fluka™
  06454-250ML
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

 

 

  • Assay:
    Min. 88.0 %
  • Density (D 20/4):
    1.190 - 1.220
  • Non-volatile matter:
    Max. 0.0001 %
  • Silver (Ag):
    Max. 0.5 ppb
  • Aluminium (Al):
    Max. 5 ppb
  • Arsenic (As):
    Max. 1 ppb
  • Gold (Au):
    Max. 1 ppb
  • Barium (Ba):
    Max. 1 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • Bismuth (Bi):
    Max. 0.5 ppb
  • Calcium (Ca):
    Max. 20 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 1 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 1 ppb
  • Iron (Fe):
    Max. 10 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Mercury (Hg):
    Max. 1 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Iridium (Ir):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 5 ppb
  • Manganese (Mn):
    Max. 0.5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 10 ppb
  • Nickel (Ni):
    Max. 2 ppb
  • Lead (Pb):
    Max. 0.5 ppb
  • Palladium (Pd):
    Max. 0.5 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 1 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Tin (Sn):
    Max. 0.5 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 1 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Uran (U):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 1 ppb
  • Zinc (Zn):
    Max. 5 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Chloride (Cl):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 0.5 ppm
  • Ammonia (NH₃):
    Max. 10 ppm
  • Acetic acid (CH₃COOH):
    Max. 0.05 %

 

 

 :
 :46.03 g/mol
 :100.4 °C (1.013 hPa)
 :4 °C
 :1.22 g/cm³ (20 °C)
 :49.5 °C
 :Ambient
 :200-579-1
 :1779
 :8,II
 :01-2119491174-37
 :GHS05
 :GHS06
 :GHS02
 :64-18-6
 :HCO₂H
 :MFCD00003297