
Caproic acid
Supplier: Roth Carl
Certificates
About this item
CAS Number:142-62-1
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00004421
:116.16 g/mol
Merck Index:13,01765
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–4.8 °C
Density:0.9276 g/cm³ (20 °C)
Flash Pt:102 °C
Storage Temperature:Ambient
EINECS:205-550-7
UN:2829
ADR:8,III
:GHS05
Frequently Bought Together