

Acetic acid glacial, anhydrous 100% (by alkalimetry), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:64-19-7
:
:H₃CCOOH
:MFCD00036152
:60.05 g/mol
:118 °C (1013 hPa)
:17 °C
:1.05 g/cm³ (20 °C)
:38.5 °C
:Ambient
:2789
:8 (3),II
:M-Clarity™ quality level = MQ300
:200-580-7
:01-2119475328-30
:GHS02
:GHS05
- Assay (Alkalimetric):≥99.8%
- Identity:Passes Test
- Color:≤10 Hazen
- Acetaldehyde:≤2 ppm
- Acetic Anhydride:≤100 ppm
- Titratable Base:≤0.0004 meq/g
- Solidification Temperature:≥16.3 °C
- Chloride (Cl):≤0.4 ppm
- Phosphate (PO₄):≤0.4 ppm
- Heavy metals (as Pb):≤0.5 ppm
- Sulfate (SO₄):≤0.4 ppm
- Ag (Silver):≤0.005 ppm
- Al (Aluminium):≤0.020 ppm
- As (Arsenic):≤0.010 ppm
- Au (Gold):≤0.010 ppm
- B (Boron):≤0.100 ppm
- Ba (Barium):≤0.010 ppm
- Be (Beryllium):≤0.005 ppm
- Bi (Bismuth):≤0.050 ppm
- Ca (Calcium):≤0.100 ppm
- Cd (Cadmium):≤0.020 ppm
- Co (Cobalt):≤0.010 ppm
- Cr (Chromium):≤0.020 ppm
- Cu (Copper):≤0.010 ppm
- Fe (Iron):≤0.050 ppm
- Ga (Gallium):≤0.050 ppm
- Ge (Germanium):≤0.020 ppm
- Hg (Mercury):≤0.005 ppm
- In (Indium):≤0.050 ppm
- K (Potassium):≤0.100 ppm
- Li (Lithium):≤0.010 ppm
- Mg (Magnesium):≤0.050 ppm
- Mn (Manganese):≤0.010 ppm
- Mo (Molybdenum):≤0.010 ppm
- Na (Sodium):≤0.200 ppm
- Ni (Nickel):≤0.020 ppm
- Pb (Lead):≤0.010 ppm
- Pt (Platinum):≤0.100 ppm
- Sn (Tin):≤0.050 ppm
- Sr (Strontium):≤0.010 ppm
- Ti (Titanium):≤0.050 ppm
- Tl (Thallium):≤0.020 ppm
- V (Vanadium):≤0.010 ppm
- Zn (Zinc):≤0.030 ppm
- Zr (Zirconium):≤0.050 ppm
- Substances Reducing Potassium Dichromate:Passes Test
- Substances Reducing Potassium Permanganate:≤20 ppm
- Evaporation Residue:≤5 ppm
- Water:≤0.2
- Dilution test:Passes Test
Frequently Bought Together