Order Entry
Belgium
ContactUsLinkComponent
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Flammable
Irritant
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99,7%
  • Evaporation residue:
    Max. 0,0005 %
  • Water:
    Max. 0,03 %
  • Pesticide analysis (Lindane/ECD):
    Max. 5 ng/l

Specification Test Results

Frequently Bought Together