

Ethyl acetate ≥99%, GPR RECTAPUR®
:141-78-6
:
:CH₃COOC₂H₅
:MFCD00009171
:88.11 g/mol
:13,03792
:77.1 °C (1013 hPa)
:–83 °C
:0.902 g/cm³ (20 °C)
:–4 °C
:Ambient
:205-500-4
:1173
:3,II
:01-2119475103-46
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Assay:Min. 99 %
- IR Spectrum:Passes test
- Acidity:Max. 0.008 meq/g
- Boiling point:76.0 - 77.5 °C
- Density (20/4):0.899 - 0.901
- Refractive index (20°C):1.371 - 1.373
- Substances coloured by H2SO4:Max. 60 APHA
- Ethanol:Max. 0.5 %
- Evaporation residue:Max. 10 ppm
- Heavy metals (as Pb):Max. 2 ppm
- Water:Max. 0.05 %
1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
spec_VWRC23880.290 spec_VWRC23880.290 spec_VWRC23880.324 spec_VWRC23880.368 spec_VWRC23880.461
Product specification
More
VWR Chemicals Big Packs and Bulks VWR® for rotary evaporation
Product Literature
Frequently Bought Together