Order Entry
Belgium
ContactUsLinkComponent
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
undefined
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Solubility:
    85.3 g/l (20 °C)
  • Melting Point:
    -83 °C
  • Molar Mass:
    88.11 g/mol
  • Boiling Point:
    77 °C (1013 hPa)
  • Vapor Pressure:
    97 hPa (20 °C)
  • Flash Point:
    -4 °C
  • Refractive Index:
    1.37
  • Explosion Limit:
    2.1 - 11.5 %(V)
  • Density:
    0.90 g/cm3 (20 °C)
  • pH:
    (H2O) no data available
  • Ignition Point:
    460 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

Related Information

Frequently Bought Together