

Ethyle acétate ≥99.6% (par GC, corrigé de la teneur en eau), ULTRA RESI-ANALYZED® pour analyse des résidus organiques, J.T.Baker®
: Avantor
:141-78-6
:
:CH₃COOC₂H₅
:MFCD00009171
:88.11 g/mol
:13,03792
:77,1 °C (1013 hPa)
:–83 °C
:0,902 g/cm³ (20 °C)
:205-500-4
:1173
:3,II
:Conditionné dans l'eau.
:–4 °C
:01-2119475103-46
:GHS02
:GHS07
:Température ambiante
- Environmentally Preferable:Safer Human and Environmental Health
- Certification environnementale:Chem21 Solvent Selection Guide recommended by default and after discussion
- For Laboratory,Research,or Manufacturing Use:= 5
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
- Assay (CH₃COOC₂H₅) (by GC, corrected for water):≥99.6 %
- Titrable Acid (µeq/g):≤0.8
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, volumetric):≤0.05 %
Frequently Bought Together