

Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
:141-78-6
:
:CH₃COOC₂H₅
:MFCD00009171
:88.11 g/mol
:13,03792
:75…78 °C (760 mmHg)
:–83.5 °C
:0.902 g/cm³
:–4 °C
:Ambient
:205-500-4
:1173
:3,II
:01-2119475103-46
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Appearance:Clear liquid
- Water:≤0.2 % (K.F.)
- Density:(20/20°C) 0.901 to 0.904
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.003 %
- Substance darkened by H2SO4:passes test
- Titrable acid:≤0.0009 meq/g
Frequently Bought Together