Order Entry
Belgium
ContactUsLinkComponent
Butyl acetate, BAKER, Laboratory Reagent, J.T.Baker®
Flammable
Irritant
Butyl acetate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 8018.1000
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Butyl acetate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 8018.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    min. 98 %
  • Acidity (as CH3COOH):
    max. 0.005 %
  • Boiling Range:
    124 - 127 °C
  • Density (g/ml) at 20°C:
    0.878 - 0.881
  • n-Butanol:
    max. 0.5 %
  • Residue after Evaporation:
    max. 0.005 %
  • Water (H2O):
    max. 0.05 %
  • Iron (Fe):
    max. 0.5
  • Heavy Metals (as Pb):
    max. 0.5

Specification Test Results

About this item

 :
 :116.16 g/mol
Merck Index:13,01534
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–76 °C
Density:0.875 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:204-658-1
UN:1123
ADR:3,III
 :GHS02
 :GHS07
CAS Number:123-86-4
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445