

Butyl acetate 99.5+% for electronic use (MOS) residue-free
Catalog # ACRO327850010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 % (Coulometric)
- GC:≥99.50 %
- Color scale:≤15 APHA
- Residue after evaporation:≤0.001 %
- Arsenic (As):≤0.01 ppm
- Barium (Ba):≤0.01 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.02 ppm
- Heavy metals (as Pb):≤50 ppm
- Acidity (CH3COOH):≤0.01 %
Specification Test Results
About this item
:
:116.16 g/mol
Merck Index:13,01534
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–78 °C
Density:0.88 g/cm³
Flash Pt:27 °C
Storage Temperature:Ambient
EINECS:204-658-1
UN:1123
ADR:3,III
:GHS07
:GHS02
CAS Number:123-86-4
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445