Acetone, Supelco®
: Merck
: 2-Propanone, DMK, Propanone, Dimethyl ketone
- :67-64-1
- :CH₃COCH₃
- :MFCD00008765
- :58.08 g/mol
- :13,00067
- :56.2 °C (1013 hPa)
- :–95.4 °C
- :0.792 g/cm³ (20 °C)
- :<–20 °C (closed cup)
- :1090
- :3,II
- :M-Clarity™ quality level = MQ100
- :Ambient
- :200-662-2
- :01-2119471330-49
Product Family Options
- Pack typePk