Acetone
Glass bottle, 2,5 L
Catalog # ROTHT906.1
Marketsource
Supplier: Roth Carl
Aller à :
Product Details & Documents
Synonymes: 2-Propanone, DMK, Propanone, Dimethyl ketone
- Numéro CAS:67-64-1
- Formule:CH₃COCH₃
- Numéro MDL:MFCD00008765
- Poids moléculaire:58.08 g/mol
- Index Merck:13,00067
- Point d'ébullition:56.2 °C (1013 hPa)
- Point de fusion:–95.4 °C
- Densité:0.792 g/cm³ (20 °C)
- Point d'éclair:<–20 °C (closed cup)
- Température de stockage:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Spécifications
Spécifications
- Pack type:Glass bottle
- Pk:2,5 L
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



