

Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
EINECS:200-662-2
UN:1090
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:<–20 °C (closed cup)
REACH:01-2119471330-49
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-95.4 °C
- Molar Mass:58.08 g/mol
- Boiling Point:56.2 °C (1013 hPa)
- Vapor Pressure:233 hPa (20 °C)
- Refractive Index:1.35868 (20 °C)
- Explosion Limit:2.6 - 12.8 %(V)
- Density:0.79 g/cm3 (20 °C)
- pH:5 - 6 (395 g/l, H2O, 20 °C)
- Ignition Point:465 °C DIN 51794
- Water Absorption:1000 g/kg
Specification Test Results
Frequently Bought Together