Watch glasses
Supplier: Brand
Jump to:
About this item
Soda-lime glass.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
Certifications: DIN 12341.
11 Options Available Below
Product Details & Documents
Soda-lime glass.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
Certifications: DIN 12341.
Product Family Options
Product Information
- ØAvailabilityPrice
- BRND150010Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges40 mmLog in for availability1 * 10 ST30,60 €
- BRND150015Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges50 mmLog in for availability1 * 10 ST30,60 €
- BRND150020Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges60 mmLog in for availability1 * 10 ST33,30 €
- BRND150030Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges70 mmLog in for availability1 * 10 ST35,50 €
- BRND150040Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges80 mmLog in for availability1 * 10 ST36,00 €
- BRND150045Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges90 mmLog in for availability1 * 10 ST40,40 €
- BRND150050Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges100 mmLog in for availability1 * 10 ST46,90 €
- BRND150060Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges120 mmLog in for availability1 * 10 ST55,10 €
- BRND150065Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges125 mmLog in for availability1 * 10 ST55,60 €
- BRND150070Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges150 mmLog in for availability1 * 10 ST69,80 €
- BRND150080Watch glass, soda-lime glass, with fire-polished edgesWatch glass, soda-lime glass, with fire-polished edges200 mmLog in for availability1 * 10 ST115,00 €