Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone

Acetone

Plastic bottle, 2,5 L
Catalog # 40289-2.5L
Flammable
Irritant
Supplier:   Honeywell Chemicals
Acetone
Catalog # 40289-2.5L
CAS Number:  67-64-1
Acetone
Acetone

Plastic bottle, 2,5 L

CAS Number:  67-64-1
Price
211,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.