Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone

Acetone

Glass bottle, 2,5 L
Catalog # FISHA/0606/17
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   FISHER CONTAINER
Acetone
Catalog # FISHA/0606/17
Supplier:  FISHER CONTAINER
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone
Acetone

Glass bottle, 2,5 L

CAS Number:  67-64-1
Price
271,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Cat. No.:
    FISHA/0606/17
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications