Order Entry
Belgium
ContactUsLinkComponent
PCR Mini Cooler with Transparent Lid
PCR Mini Cooler with Transparent Lid
Catalog # 211-3269
Supplier:  Brand
undefined
PCR Mini Cooler with Transparent Lid
Catalog # 211-3269
Supplier:  Brand

Specifications

  • For:
    PCR tubes 0,2 ml single, 8-strips, 12-strips and 96-well PCR plates
  • Pk:
    2

Specifications

About this item

PP

  • The PCR Mini cooler keeps samples at 4 °C for about 3 hours
  • The insulating gel changes from violet to pink at 7 °C
  • Suitable for 0,2 ml single vessels, 8-strips, 12-strips as well as 96-well PCR plates

To protect the samples prior to warming.