Microplates, 96-well
96-well microplate with U-bottom, 300 μl
Catalog # 391-5708
Supplier: Brand
Jump to:
Product Details & Documents
PP, round bottom wells, non sterile.
- Compatible with virtually all leading microplate centrifuges
- Raised well rims minimise cross-contamination
- Plates can be sealed using self-adhesive films, such as DMSO-resistant sealing film (cross-cut) with alphanumeric coding
For volumes up to 300 µl.
Documents
Brand Deep-well Plates
Product Literature
Specifications
- Description:96-well microplate with U-bottom, 300 μl