Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone, BAKER, J.T.Baker®

Acetone, BAKER, J.T.Baker®

Flammable
Irritant
   Sustainable Options Available
Supplier:   Avantor
Acetone, BAKER, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
Acetone, BAKER, J.T.Baker®

6 Options Available Below

Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications for Product Family

Specification Test Results

  • Acidity or Alkalinity:
    passes test
  • Appearance of solution:
    passes test
  • Identification:
    passes test
  • Matter Insoluble in Water:
    passes test
  • Reducing Substances:
    passes test
  • Relative Density:
    0.790-0.793
  • Residue on Evaporation:
    max. 50 ppm
  • Water (H2O):
    max. 3 g/l
  • Additional Specification(s)::
    -
  • Assay (by GC):
    min. 99,0 %
  • Related Substances::
    -
  • Benzene:
    max. 2 ppm (v/v)
  • Methanol (CH3OH):
    max. 0,05 % (v/v)
  • Other Impurities:
    passes test
  • 2-Propanol (CH3CHOHCH3):
    max. 0,05 % (v/v)

Specifications

Specifications

  • Catalog No:
  • 8003.1000
  • 8003.2500
  • 8003.5000
  • 8003.9010
  • 8003.9025
  • 8003.9200ST
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,09 €
    41,09 €
    1 * 1 L
       41,09 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    8003.1000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 83,80 €
    83,80 €
    1 * 2,5 L
       83,80 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    8003.2500
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 153,32 €
    153,32 €
    1 * 5 L
       153,32 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    8003.5000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Plastic container
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 276,38 €
    276,38 €
    1 * 10 L
       276,38 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    8003.9010
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 287,95 €
    287,95 €
    1 * 25 L
       287,95 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    8003.9025
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 200 L
    Log in for availability
    Log in for availability
    1 * 200 L
    1 * 200 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    8003.9200ST
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.