Tubing, silicone
Supplier: Brand
Certificates
About this item
Silicone, translucent.
- Excellent temperature resistance
- Autoclavable (121 °C)
- Limited chemical resistance when used with concentrated acids
For laboratory application.
Frequently Bought Together