Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone ≥99.5% for HPLC

Acetone ≥99.5% for HPLC

Flammable
Irritant
   Sustainable Options Available
Acetone ≥99.5% for HPLC
   Sustainable Options Available
Certificates
Acetone ≥99.5% for HPLC

2 Options Available Below

Product Details & Documents

Purity: 99.5+% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3590 Appearance: Colourless Odour: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 635680 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765

Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:14,66
  • Boiling Pt:56 °C
  • Melting Pt:–94 °C
  • Density:0.791
  • Flash Pt:–17 °C (1 °F)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 22928.K2
  • 22928.M1
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk Info
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 76,00 €
    76,00 €
    1 * 1 L
       76,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    22928.K2
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 4x1 L
    4
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 368,00 €
    368,00 €
    1 * 4 ST
       368,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    22928.M1
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.