Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone 99+% for spectroscopy

Acetone 99+% for spectroscopy

 
 
    
Acetone 99+% for spectroscopy
    
 
Acetone 99+% for spectroscopy
Acetone 99+% for spectroscopy

 

 
 :  2-Propanone, DMK, Propanone, Dimethyl ketone
  •  :67-64-1
  •  :CH₃COCH₃
  •  :MFCD00008765
  •  :58.08 g/mol
  •  :13,00067
  •  :56.2 °C (1013 hPa)
  •  :–95 °C
  •  :0.79 g/cm³
  •  :–20 °C (closed cup)
  •  :Ambient
  •  :200-662-2
  •  :1090
  •  :3,II
  •  :01-2119471330-49

Documents

 

 

  • Appearance:
    Clear liquid
  • Water:
    ≤0.5 % (Coulometric)
  • GC:
    ≥99.0 %
  • UV:
    at 330 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %

 

 

  • Catalog No:
  • ACRO167645000
  • ACRO167640025
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

 

Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle
    500 mL
       
       
    1 * 500 mL
    1 * 500 mL - 34,50 €
    34,50 €
    1 * 500 mL
       34,50 €

     
      Safer Human and Environmental Health
     
    Specifications
    Cat. No.
    ACRO167645000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Glass bottle
    2,5 L
       
       
    1 * 2,5 L
    1 * 2,5 L - 96,00 €
    96,00 €
    1 * 2,5 L
       96,00 €

     
      Safer Human and Environmental Health
     
    Specifications
    Cat. No.
    ACRO167640025
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.