Orders LinkContactUsLinkComponent
Funnel stands
Funnel stands
Supplier:   Brand
Funnel stands
Supplier:  Brand
Certificates
Funnel stands
2 Options Available Below
Product Details & Documents
PP plates for holding two or four funnels with top outer-Ø of 50 to 120 mm.
  • With aluminum support rod (Ø×L: 12,7×595 mm)
  • Adjustable funnel height
Documents
Product Family Options
Product Information
  • L×W
    For
    Availability
    Price
  • 519-2711
    Plates with aluminum support rod (Ø×L: 12,7×595 mm)
    Plates with aluminum support rod (Ø×L: 12,7×595 mm)
    250×140 mm
    2 funnels
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 198,00 €
    198,00 €
    1 * 1 ST
       198,00 €
  • BRND74511
    Plates with aluminum support rod (Ø×L: 12,7×595 mm)
    Plates with aluminum support rod (Ø×L: 12,7×595 mm)
    450×140 mm
    4 funnels
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 225,00 €
    225,00 €
    1 * 1 ST
       225,00 €