Orders LinkContactUsLinkComponent
UV microplates, Nunc™
UV microplates, Nunc™
96-well plate, UV-transparent base, Clear, 342 μl, 96
Catalog # 735-0253
   Low Manufacturing Impact
UV microplates, Nunc™
Catalog # 735-0253
   Low Manufacturing Impact
UV microplates, Nunc™
UV microplates, Nunc™
96-well plate, UV-transparent base, Clear, 342 μl, 96
Price
780,00 €
Log in for availability
1 * 40 ST
Product Details & Documents
Perform DNA and protein quantitation in 260 nm and 280 nm range with Thermo Scientific™ Nunc™ 96-Well UV microplates. The 96-Well format allows for a higher throughput of sample screening compared to cuvettes.
  • Non sterile
  • Flat bottom (F-well)
Documents
Specifications
  • No. of wells:
    96
  • Colour:
    Clear
  • Description:
    96-well plate, UV-transparent base
  • Well volume:
    342 μl
  • Cat. No.:
    735-0253
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact