Orders LinkContactUsLinkComponent
Stopcocks, Nalgene®
Stopcocks, Nalgene®
   Sustainable Options Available
Stopcocks, Nalgene®
   Sustainable Options Available
Certificates
Stopcocks, Nalgene®
2 Options Available Below
Product Details & Documents
Polypropylene with PTFE plug.
  • Autoclavable
  • Very good chemical resistance
  • Serrated tubulation on each end accepts 1/2" through 5/16" tubing
Permit reliable, accurate liquid flow control in liquid handling systems. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.
Specifications for Product Family
Specifications
  • Catalog No:
  • 229-0016
  • 229-0017
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Hole Ø
    For tube I-Ø
    Availability
    Price
  • 2 mm
    6,35 - 9,53 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,70 €
    67,70 €
    1 * 1 ST
       67,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    229-0016
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 4 mm
    6,35 - 9,53 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,10 €
    66,10 €
    1 * 1 ST
       66,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    229-0017
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact