Supelco® Organic Reference Standard, 3-Octanone
46214-5ML
: Merck
:
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:106-68-3
- Description:3-Octanone
- Storage conditions:2...30 °C
- Hazard class description:H226 - H315
- Shipping conditions:STD
- MDL No.:MFCD00009515
- M-Clarity quality level:MQ100
- Synonym(s):Ethyl amyl ketone; Ethyl pentyl ketone
- Formula:CH₃(CH₂)₄COC₂H₅
- Molecular weight:128,21
- Density:0,822 g/ml at 25 °C (lit.)
- Flash point:51 °C - Closed cup
- Boiling point:167 - 168 °C (lit.)
- GHS signal: GHS02
: P210 - P233 - P240 - P241 - P242 - P303 + P361 + P353