
Patton-reeders reagent (Calconcarboxylic acid dihydrate) indicator for metal titration, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:3737-95-9
Formula:C₂₁H₁₄N₂O₇S·2H₂O
MDL Number:MFCD00004078
:474.45 g/mol
:M-Clarity™ quality level = MQ200
EINECS:223-117-0
:GHS07
Specification Test Results
- Appearance of solution:passes test
- Water (according to Karl Fischer):≤ 9%
- Suitability as indicator (for metal titration):passes test
- Bulk Density:400 kg/m3
Specification Test Results
Related Information
MERC1.04595.0005_TDS_EN
Product specification
Merck Brighter titration Titripur®
Product Literature
msds_104595
SDS
Frequently Bought Together