Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Catalog # HYPETS-48914
CAS Number:  
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Catalog # HYPETS-48914
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :199.25 g/mol
Boiling Pt:130 °C (1013 hPa)
Density:1,07 g/cm³ (20 °C)
EINECS:246-331-6
 :GHS02
 :GHS07
CAS Number:24589-78-4
Formula:C₆H₁₂F₃NOSi
MDL Number:MFCD00000411
Boiling Pt:130 °C (1013 hPa)