Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pyridine

Pyridine

Glass bottle, 50 mL
Catalog # HYPETS-27530
Flammable
Irritant
Pyridine
Catalog # HYPETS-27530
CAS Number:  110-86-1
Pyridine
Pyridine

Glass bottle, 50 mL

CAS Number:  110-86-1
Price
149,00 €
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications