Orthophosphoric acid 85%
Product Details & Documents
Synonyms: Phosphoric acid
- CAS Number:7664-38-2
- Formula:H₃PO₄
- MDL Number:MFCD00011340
- Molecular Weight:98 g/mol
- Melting Pt:28 °C
- Density:1.71…1.87 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:231-633-2
- UN:1805
- ADR:8,III
Documents
Certificates
Search by Lot Numberspec_VWRS5992.9020 spec_VWRS5992.9020 spec_VWRS5992.9020



