Sodium hydroxide 5 mol/l (5 N) in aqueous solution 5 mol/l (5 N) in aqueous solution, Titrisol®, Supelco®
Catalog # 1.09913.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.185 g/cm3 (20 °C)
- pH:14 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
Synonyms: Caustic soda
:
:40 g/mol
Boiling Pt:1390
EINECS:215-185-5
UN:1824
:GHS05
Storage Temperature:Ambient
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548
Melting Pt:323
Density:≤1.11 g/cm³ (20 °C)
ADR:8
Merck Index:14,08627
:M-Clarity™ quality level = MQ100