Order Entry
Belgium
ContactUsLinkComponent
Disodium hydrogen phosphate, anhydrous ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Disodium hydrogen phosphate, anhydrous ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Disodium hydrogen phosphate, anhydrous ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.01%
  • Assay (Na₂HPO₄) (by acidimetry):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.01 %
  • Loss on Drying at 105°C:
    ≤0.2 %
  • pH of 5% Solution at 25°C:
    8.7 - 9.3
  • Chloride (Cl):
    ≤0.002 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Iron (Fe):
    ≤0.002 %

Specification Test Results

Related Information

Frequently Bought Together