Order Entry
Belgium
ContactUsLinkComponent
Palladium(II) acetate for synthesis, Sigma-Aldrich®
Corrosive
Palladium(II) acetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Palladium(II) acetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    205 °C
  • Molar Mass:
    224.5 g/mol
  • pH:
    2 - 3 (H2O, 20 °C) (aqueous suspension)

Specification Test Results

Related Information

Frequently Bought Together