Order Entry
Belgium
ContactUsLinkComponent
Pineapple Bromelain (from Stem)
Pineapple Bromelain (from Stem)
Catalog # A1548.0500
Supplier:  PanReac AppliChem
CAS Number:  
Pineapple Bromelain (from Stem)
Catalog # A1548.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • CAS No.:
    37189-34-7
  • Source:
    Stem
  • Species:
    Pineapple
  • Enzyme name:
    Bromelain

Specifications

About this item