Organic reference standard, Bromochloromethane 1000 µg/ml in methanol
Catalog # CMPLD1A7.1K.ML.1
Supplier: CUSTOM MADE CHEMICALS LAB
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 Ampoul
- CAS No.:74-97-5
- Description:Bromochloromethane 1,000 µg/ml in methanol
- Formula:CH₂BrCl
- Description:Bromochloromethane