5-Chloro-3-methyl-1-phenyl-1H-pyrazole
Catalog # MAYBBTB08985.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.65 g/mol
CAS Number:1131-17-5
Formula:C₁₀H₉ClN₂
MDL Number:MFCD00067853