0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Acetonitrile
Acetonitrile
Acetonitrile
:
Roth Carl
Acetonitrile
:
Roth Carl
:
75-05-8
:
:
H₃CCN
:
MFCD00001878
:
41.05 g/mol
:
13,00071
:
81.6 °C (1013 hPa)
:
–45.7 °C
:
0.782 g/cm³ (20 °C)
:
2 °C (closed cup)
:
Ambient
:
200-835-2
:
1648
:
3,II
:
01-2119471307-38
:
GHS02
:
GHS07
Frequently Bought Together
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
: Merck
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection