

Acetonitrile ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:13,00071
:81.6 °C (1013 hPa)
:–45,7 °C
:0.782 g/cm³ (20 °C)
:5 °C (closed cup)
:Ambient
:200-835-2
:1648
:3,II
:01-2119471307-38
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Assay (on anhydrous substance):Min. 99.5 %
- IR Spectrum:Passes test
- Solution (100 g/l):Passes test
- Acidity:Max. 0.0002 meq/g
- Alkalinity:Max. 0.0006 meq/g
- Boiling point:80 - 82 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.781 - 0.784
- Density (20/20):0.782 - 0.785
- Refractive index (20°C):1.343 - 1.345
- Evaporation residue:Max. 10 ppm
- Water:Max. 0.15 %
- CN (Cyanide):Max. 50 ppm
- Cd (Cadmium):Max. 0.02 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cu (Copper):Max. 0.04 ppm
- Fe (Iron):Max. 0.05 ppm
- Mn (Manganese):Max. 0.01 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.05 ppm
- Zn (Zinc):Max. 0.1 ppm
- Transmittance (255-420 nm):Min. 98 %
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Frequently Bought Together