

Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45,7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay:Min. 99 %
- Refractive index (20°C):1.343 - 1.345
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
spec_VWRC20070.291 spec_VWRC20070.291 spec_VWRC20070.325 spec_VWRC20070.325 spec_VWRC20070.462
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together