
1-(2-Pyridylazo)-2-naphthol (PAN) 98%, pure indicator grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:85-85-8
:
Formula:C₁₅H₁₁N₃O
MDL Number:MFCD00004071
:249.27 g/mol
Boiling Pt:412 °C (1013 hPa)
Melting Pt:138…141 °C
Storage Temperature:Ambient
EINECS:201-637-9
:GHS07
Specification Test Results
- Appearance:Orange to red-brown to brown Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
- Suitability:(as complexom. indicator) Passes test
Specification Test Results
Frequently Bought Together