Order Entry
Belgium
ContactUsLinkComponent
Pesticide reference standard, 2,4,6-Trichlorophenol 1,000 µg/ml in methanol
Pesticide reference standard, 2,4,6-Trichlorophenol 1,000 µg/ml in methanol
Supplier:  Restek
Certificates
Pesticide reference standard, 2,4,6-Trichlorophenol 1,000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    88-06-2
  • Formula:
    C₆H₃Cl₃O
  • Description:
    2,4,6-Trichlorophenol

Specifications

Frequently Bought Together