Pesticide reference standard, 2,4,5-Trichlorophenol 1,000 µg/ml in methanol
Supplier: Restek
Certificates
About this item
Specifications
- CAS No.:95-95-4
- Formula:C₆H₃Cl₃O
- Description:2,4,5-Trichlorophenol
Specifications
Frequently Bought Together