Acetonitrile, LiChrosolv®, Isocratic Grade for liquid chromatography, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
EINECS:200-835-2
UN:1648
ADR:3,
:M-Clarity™ quality level = MQ100
Flash Pt:2 °C (closed cup)
REACH:01-2119471307-38
:GHS02
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Related Information
Certificates
Search by Lot NumberMerck Pharma Segment Brochure
Product Literature
msds_114291
SDS
ProductPage_114291
Product specification
Frequently Bought Together