

Acetonitrile, anhydrous (max. 0.001 % H₂O)
: Merck
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:13,00071
:81.6 °C (1013 hPa)
:–45.7 °C
:0.782 g/cm³ (20 °C)
:200-835-2
:1648
:3,
:M-Clarity™ quality level = MQ200
:2 °C (closed cup)
:01-2119471307-38
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Solubility:(20 °C) soluble
- Melting Point:-45.7 °C
- Molar Mass:41.05 g/mol
- Boiling Point:81.6 °C (1013 hPa)
- Vapor Pressure:97 hPa (20 °C)
- Flash Point:2 °C
- Refractive Index:1.34
- Explosion Limit:3.0 - 17 %(V)
- Density:0.786 g/cm3 (20 °C)
- pH:(H2O) no data available
- Ignition Point:524 °C
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together