Order Entry
Belgium
ContactUsLinkComponent
Pesticide reference standard, 2,6-Dichlorophenol 1,000 µg/ml in methanol
Pesticide reference standard, 2,6-Dichlorophenol 1,000 µg/ml in methanol
Supplier:  Restek
Certificates
Pesticide reference standard, 2,6-Dichlorophenol 1,000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    87-65-0
  • Formula:
    C₆H₄Cl₂O
  • Description:
    2,6-Dichlorophenol

Specifications

Frequently Bought Together