Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Acetonitrile, SupraSolv® for gas chromatography, Supelco®
 :  Merck
    
 
undefined
 
 
Acetonitrile, SupraSolv® for gas chromatography, Supelco®
 :  Merck
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Solubility:
    (20 C) soluble
  • Melting Point:
    -45.7 C
  • Molar Mass:
    41.05
  • Boiling Point:
    81.6 C
  • Vapor Pressure:
    97 hPA
  • Flash Point:
    2 C
  • Refractive Index:
    1.34
  • Explosion Limit:
    3.0 - 17 %
  • Density:
    0.786 g/cm3
  • pH:
    (H20) N/A
  • Ignition Point:
    524 C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

 

Frequently Bought Together